Learn More
ACROBiosystems Human IL-34 Protein, His Tag
Supplier:  ACROBiosystems IL4H52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-34, His Tag (IL4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asn 21 - Pro 242 (Accession # NP_689669.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.1 kDa.
Catalog No. 50-205-4805