Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-35 ELISA Kit

Catalog No. EEL067
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL067 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL067 Supplier Invitrogen™ Supplier No. EEL067
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human IL-35 ELISA quantitates IL-35 in serum, plasma, and other biological fluids The Human IL-35 (Interleukin 35) solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL-35, or Interleukin 35 is a cytokine belonging to the IL12 family, that is produced by regulatory T cells. It is a dimeric protein composed of IL-12-alpha and IL-27-beta chains. Members of this class of cytokines are important in influencing T helper cell differentiation and supressing inflammatory response. IL-35 is known to induce proliferation of regulatory T cells, and reduce the activity of helper Th17 cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29459
Assay Range 15.63-1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family IL-12
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CLMF; CLMF p35; cytokine receptor; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; EBI3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; epstein-Barr virus-induced gene 3 protein; IL-12 subunit p35; IL-12, subunit p35; IL12A; IL-12A; IL12p35; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; il35; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; interleukin-12 subunit alpha; Interleukin-27 subunit beta; Interleukin-35; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35CLMF; CLMF p35; cytokine receptor; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; EBI3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; epstein-Barr virus-induced gene 3 protein; IL-12 subunit p35; IL-12, subunit p35; IL12A; IL-12A; IL12p35; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; il35; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; interleukin-12 subunit alpha; Interleukin-27 subunit beta; Interleukin-35; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35;
Gene ID (Entrez) 3592
Gene Symbol IL12A
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 100 μL, Serum 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target IL-35
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.