Learn More
ACROBiosystems Human IL-35 R His Tag
Supplier:  ACROBiosystems ILRH52H850UG
Encompass_Preferred
Human IL-35 R His Tag (ILR-H52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Glu 619 & Lys 24 - Asn 622 (Accession # P40189-1(gp130) & Q99665-1(IL12Rb2)).
Catalog No. 50-253-4574