Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human IL-35 Recombinant Protein, PeproTech®

Catalog No. 2003750UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
2 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
200-37-50UG 50 μg
200-37-10UG 10 μg
200-37-2UG 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 200-37-50UG Supplier Gibco™ Supplier No. 2003750UG
  • $714.00
    $535.65 / Each of 1

    Save $178.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

200-37-100UG will be provided as 2 x 50 μg (200-37-50UG). 200-37-1MG will be provided as 20 x 50 μg (200-37-50UG). 200-37-250UG will be provided as 5 x 50 μg (200-37-50UG). 200-37-500UG will be provided as 5 x 100 μg (200-37-50UG). Recombinant Recombinant Human IL-35 expressed in HEK293 cells contains 406 amino acid residues and has a calculated molecular weight of 45.8 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.

IL-35, or Interleukin 35 is a cytokine belonging to the IL12 family, that is produced by regulatory T cells. It is a dimeric protein composed of IL-12-alpha and IL-27-beta chains. Members of this class of cytokines are important in influencing T helper cell differentiation and supressing inflammatory response. IL-35 is known to induce proliferation of regulatory T cells, and reduce the activity of helper Th17 cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29459
For Use With (Application) Bioactivity, Control
Formulation Protein with no preservative
Gene ID (Entrez) 3592
Molecular Weight (g/mol) 45.8 kDa
Name Human IL-35
Purification Method Purified
Quantity 50 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias CLMF; CLMF p35; cytokine receptor; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; EBI3; EBV-induced gene 3 protein; Epstein-Barr virus induced 3; Epstein-Barr virus induced gene 3; epstein-Barr virus-induced gene 3 protein; IL-12 subunit p35; IL-12, subunit p35; IL12A; IL-12A; IL12p35; IL27 subunit; IL-27 subunit beta; IL27B; IL-27B; il35; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin-12 alpha chain; interleukin-12 subunit alpha; Interleukin-27 subunit beta; Interleukin-35; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NKSF1; P35
Common Name IL-35
Gene Symbol IL12A
Biological Activity Measured by its ability to inhibit proliferative activity of PBMCs.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Alpha Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS Beta Subunit: RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK
Content And Storage -20°C
Activity Measured by its ability to inhibit proliferative activity of PBMCs.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.