Learn More
Sino Biological Human IL-4 ELISpot Kit (ALP) 1 Kit (96 Tests)
Catalog No. 5000696236
Encompass_Preferred
The kit applies for the detection of human IL-4 at the cellular level by counting IL-4-secreting cells.
Catalog No. 50-006-96236 Supplier Sino Biological Supplier No. EK11846A1
The kit applies for the detection of human IL-4 at the cellular level by counting IL-4-secreting cells.