Learn More
ACROBiosystems ACROBiosystems Human IL-4 R alpha / CD124 Protein, His Tag
Supplier:  ACROBiosystems ILRH52211MG
Encompass_Preferred
Recombinant Protein;1MG;Human IL-4 R alpha, His Tag (ILR-H5221) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # NP_000409.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.6 kDa. The protein migrates as 36-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-4 R alpha, His Tag (Cat. No. ILR-H5221) at 5μg/mL (100 μL/well) can bind Biotinylated Human IL-4, Avitag,His Tag (Cat. No. IL4-H82E0) with a linear range of 2-16 ng/mL (QC tested).;Serial dilutions of Monoclonal IL-4R/CD124 Neutralizing Antibody were added into Human IL-4 R alpha, His Tag (Cat. No. ILR-H5221): Biotinylated Human IL-4, Avitag,His Tag (Cat. No. IL4-H82E0) binding reactions. The half maximal inhibitory concentration (IC50) is 0.505 μg/mL (Routinely tested).
Catalog No. 50-210-6682