Learn More
ACROBiosystems Human IL-4 R alpha IL-13 R a
Catalog No. 502534576
Encompass_Preferred
Human IL-4 R alpha IL-13 R alpha 1 Protein Fc TagFc Tag (ILR-H525b) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 Gly 22 - Thr 343 (Accession # P24394-1 NP_001551.1).
Catalog No. 50-253-4576 Supplier ACROBiosystems Supplier No. ILRH525B1MG
Human IL-4 R alpha IL-13 R alpha 1 Protein Fc TagFc Tag (ILR-H525b) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 Gly 22 - Thr 343 (Accession # P24394-1 NP_001551.1).