Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-5 ELISpot Kit

Catalog No. XEL205
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
XEL205 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. XEL205 Supplier R&D Systems™ Supplier No. XEL205
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly sensitive, microplate-based assays for the detection of cytokine secreting cells

  • Detect and quantitate individual cells secreting
  • High sensitivity - ELISpot assays can measure responses with frequencies well below 1 in 100,000 cells
  • No in vitro expansion of cells required
  • High-throughput - ELISpot assays use only a small number of primary cells

Specifications

Assay Quantitative Sandwich ELISA
Immunoassay Kit Format 96-well PVDF-backed Microplate
Sample Type Whole Cells
Gene ID (Entrez)
  • 3567 (Human)
  • 16191 (Mouse)
  • 24497 (Rat)
  • 397409 (Porcine)
  • 280825 (Bovine)
  • 403790 (Canine)
  • 493803 (Feline)
Kit Contents
  • Microplate
  • Biotinylated Detection Antibody
  • Streptavidin conjugated to Alkaline Phosphatase
  • Dilution Buffers
  • Wash Buffer Concentrate
  • BCIP/NBT Chromogen
  • Positive Control
Quantity 1 Pack
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Total Assay Time 3 hr. 15 min.s to 4 hr. 30 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.