Learn More
Human IL-5 ELISPOT Pair, BD
Catalog No. BDB551886 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 x 96-well Plates
Catalog No. Quantity
BDB551886 5 x 96-well Plates
1 options
Catalog No. BDB551886 Supplier BD Biosciences Supplier No. 551886
Includes
Human IL-5 ELISPOT Capture Antibody (no azide/low endotoxin), Human IL-5 ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations
Convenient set of unlabelled anti-IL-5 capture antibody and a biotinylated anti-IL-5 detection antibody
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 x 96-well Plates