Learn More
ACROBiosystems Human IL-5 R alpha 200ug
Catalog No. 502534600
Encompass_Preferred
Biotinylated Human IL-5 R alpha Protein AvitagFc Tag (ILA-H82F5) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Trp 342 (Accession # Q01344-1).
Catalog No. 50-253-4600 Supplier ACROBiosystems Supplier No. ILAH82F5200UG
Biotinylated Human IL-5 R alpha Protein AvitagFc Tag (ILA-H82F5) is expressed from human 293 cells (HEK293). It contains AA Asp 21 - Trp 342 (Accession # Q01344-1).