Learn More
Kingfisher Biotech Inc FNDC5 ELISA - Less than or equal to 10 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY2095H004
Encompass_Preferred
The Human/Monkey FNDC5 (Irisin) ELISA contains capture antibody, standard, and detection antibody for development of a Human/Monkey FNDC5 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. This ELISA will measure FNDC5 (Irisin) of over 50 species, including Human, Monkey, and Dolphin since the protein sequence is 100% identical. Contact Kingfisher Biotech, Inc. for species list. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5686