Learn More
TRIBIOSCIENCE INC Human IL-6 ELISA, 1 x 96-well plate
SDP
Catalog No. NC3454591
Encompass_Preferred
Tribioscience’s Human IL-6 ELISA is designed to quantitatively detect human IL-6 levels in in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 3 to 800 pg/mL.
Catalog No. NC3454591 Supplier TRIBIOSCIENCE INC Supplier No. TBS3223
Tribioscience’s Human IL-6 ELISA is designed to quantitatively detect human IL-6 levels in in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 3 to 800 pg/mL.