Learn More
Raybiotech Inc Human IL-6 ELISA Kit
Supplier:  Raybiotech Inc ELHIL6CL1
Encompass_Preferred
For cell and tissue lysate samples. Sensitivity: 3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human IL-6.
Catalog No. 50-194-9455