Learn More
Abcam Human IL-6 ELISA Kit, 1 x 96 Tests
Catalog No. NC1164336
Encompass_Preferred
Human IL-6 ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human interleukin-6 (IL-6) with a sensitivity of 1.6 pg/mL. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450nm readout; works on any plate reader - Different formats for different needs: 10x 96-well plates for bulk orders and 384-well for higher throughput - Broad sample compatibility – including CSF - Cited in over 210 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1164336 Supplier Abcam Supplier No. AB1780131X96TESTS
Human IL-6 ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human interleukin-6 (IL-6) with a sensitivity of 1.6 pg/mL. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450nm readout; works on any plate reader - Different formats for different needs: 10x 96-well plates for bulk orders and 384-well for higher throughput - Broad sample compatibility – including CSF - Cited in over 210 publications
The product is subject to the following: Abcam Restricted Use Statement