Learn More
FUJIFILM BIOSCIENCES INC Human IL-6 , Recombinant Protein, Shenandoah Biotechnology, 20 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1001020UG
Encompass_Preferred
Interleukin 6 (IL-6) is an important pro-inflammatory and anti-inflammatory cytokine expressed by T cells, macrophages, and muscle cells.  IL-6 signals through a receptor complex containing two receptors, IL-6RA and gp130.  IL-6 has an important function in promoting fever and can serve to stimulate an immune response to trauma.  IL-6 is often used for growth of hybridoma cell lines.  Human IL-6 is active on mouse and rat cells.  
Catalog No. 50-990-625