Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems Human IL-6 Quantikine ELISA Kit

Catalog No. D6050B
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D6050B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. D6050B Supplier R&D Systems Supplier No. D6050B
Add to Cart
Add to Cart
This item is not returnable. View return policy

For measuring human IL-6 in cell culture supernates, serum, plasma, and urine

The Quantikine Human IL-6 Immunoassay is a 4.5 hour solid phase immunoassay designed to measure human IL-6 in cell culture supernates, serum, plasma, and urine. It contains recombinant human IL-6 and antibodies raised against the recombinant protein. Natural human IL-6 showed dose-response curves that were parallel to the standard curves obtained using the Quantikine kit standards, indicating that this kit can be used to determine relative levels of natural human IL-6.

Specifications

Assay Range 3.13 - 300 pg/mL
Assay Sensitivity 0.626 pg/mL
Sample Type Cell Culture Supernatant, Serum, Plasma (EDTA), Plasma (Heparin), Urine
Specificity Natural and recombinant human IL-6.
Gene ID (Entrez) 3569
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.