Learn More
ACROBiosystems Biotinylated Human IL-6 R alpha, Fc,Avitag (ILR-H82F9) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 365 (Accession # P08887-1).
Supplier:  ACROBiosystems ILRH82F925UG
Encompass_Preferred
Interleukin 6 receptor (IL6R) is also known as CD126 (Cluster of Differentiation 126) , is a potent pleiotropic cytokine that regulates cell growth and differentiation of various tissues, and is known particularly for its role in the immune response and acute phase reactions. IL6R is a protein complex consisting of a IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. IL6R also denotes the human gene encoding this subunit. Alternatively spliced transcript variants encoding distinct isoforms have been reported. IL6R subunit also shared by many other cytokines. The soluble form of IL6R arises from proteolytic cleavage of membrane-bound IL6Rα, and acts agonistically by making the IL6 ligand accessible to the signal transducer gp130.
Catalog No. 50-223-1008