Learn More
Bon Opus Biosciences Recombinant Human IL-6R (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107750UG
Encompass_Preferred
Recombinant Human IL-6R (C-hFc) is produced by Human Cells. The target gene encoding Leu20-Ser359 is expressed with a C-hFc tag. Accession# P08887-1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1077
Catalog No. 50-217-2675