Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human IL-7 Protein (E. coli), Recombinant, Premium Grade, 50 μg, Lyophilized

Catalog No. NC3815913
Encompass_Preferred

Recombinant human interleukin-7 (IL-7) expressed in E. coli and supplied as a premium-grade, lyophilized powder. Designed for bioactivity assays, cell culture stimulation, and early-stage cell therapy development; calibrated to the WHO IL-7 standard for consistent activity measurements.

  • Stimulates proliferation of PHA-P-activated human PBMC.
  • Specific activity greater than 1.00x10^8 IU/mg (WHO calibrated).
  • Purity >95% by SDS-PAGE.
  • Contains amino acids Asp 26-His 177; no affinity tag.
  • Formulated with trehalose and sterile filtered prior to lyophilization.
  • Store lyophilized at -20 °C or lower; avoid repeated freeze-thaw after reconstitution.

Catalog No. NC3815913 Supplier ACROBiosystems Supplier No. IL7H511650UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-7 (IL-7) expressed in E. coli and supplied as a premium-grade, lyophilized powder. Designed for bioactivity assays, cell culture stimulation, and early-stage cell therapy development; calibrated to the WHO IL-7 standard for consistent activity measurements.

  • Stimulates proliferation of PHA-P-activated human PBMC.
  • Specific activity greater than 1.00x10^8 IU/mg (WHO calibrated).
  • Purity >95% by SDS-PAGE.
  • Contains amino acids Asp 26-His 177; no affinity tag.
  • Formulated with trehalose and sterile filtered prior to lyophilization.
  • Store lyophilized at -20 °C or lower; avoid repeated freeze-thaw after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.