Learn More
Raybiotech Inc Human IL-7 R alpha/CD127 ELISA Kit
Supplier:  Raybiotech Inc ELHIL7RA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.14 ng/ml.. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human IL-7 R alpha.
Catalog No. 50-194-9458