Learn More
ACROBiosystems Human IL-7 R alpha&TSLP R Heterodimer Protein, Fc Tag&Fc Tag (MALS verified)
Supplier:  ACROBiosystems ILRH5255100UG
Encompass_Preferred
Recombinant Protein;100UG;Human IL-7 RA&TSLP R Heterodimer Protein, Fc Tag&Fc Tag (ILR-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Asp 239 (IL-7 RA) & Gln 23 - Lys 231 (TSLP R) (Accession # P16871-1 (IL-7RA) & Q9HC73-1 (TSLPR)).;Human IL-7 RA&TSLP R Heterodimer Protein, Fc Tag&Fc Tag is produced by co-expression of IL-7 RA and TSLP R, has a calculated MW of 51.4 kDa (IL-7 RA) and 50.4 kDa (TSLP R). Subunit IL-7 RA is fused with a human IgG1 Fc fragment at the C-terminus and subunit TSLP R is fused with a human IgG1 Fc fragment at the C-terminus.
Catalog No. 50-205-4804