Learn More
Biotang Inc Human IL-8 / CXCL8 Protein (aa 23-99, Fc Tag),50ug, expressed in human cells.the 77 amino acid endothelial-cell derived form ofthe mature human IL8 (NP_000575.1) (Ala 23-Ser 99) was fused with the Fc region of human IgG1 at the N-terminus.
Supplier:  Biotang Inc RPR9320
Encompass_Preferred
Human IL-8 / CXCL8 Protein (aa 23-99, Fc Tag),50ug, expressed in human cells.the 77 amino acid endothelial-cell derived form ofthe mature human IL8 (NP_000575.1) (Ala 23-Ser 99) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-751-5544