Learn More
Biotang Inc Human IL-8 / CXCL8 Protein (aa 28-99, Fc Tag), 50ug, expressed in human cells. the 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) was fused with the Fc region of human IgG1 at the N-terminus.
Supplier:  Biotang Inc RPR9322
Encompass_Preferred
Human IL-8 / CXCL8 Protein (aa 28-99, Fc Tag), 50ug, expressed in human cells. the 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-751-5543