Learn More
R&D Systems™ Human IL-8/CXCL8 Quantikine HS ELISA Kit
Description
Specifications
Specifications
| Assay Range | 1.00 - 64 pg/mL |
| Assay Sensitivity | 0.4 pg/mL |
| Conjugate | HRP |
| Sample Type | Plasma (EDTA), Heparin Plasma, Serum, Serum |
| Specificity | Natural and Recombinant Human IL-8 |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3576 |
| Quantity | One 96-well plate |
| Sample Volume | Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.