Learn More
Abcam Human IL-8 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2140301X96TESTS
Encompass_Preferred
Human IL-8 ELISA kit is a single-wash, 90-minute SimpleStep ELISA used to quantify human interleukin-8 (IL-8) with a sensitivity of 1.8 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma, cell culture supernatants, and cerebrospinal fluid - Cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2168055