Learn More
Sino Biological Recombinant Anti-IL-10 Antibody (Biotin), Rabbit Monoclonal
Supplier:  Sino Biological 10947R128B100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL10 / IL-10 / Interleukin-10 (rh IL10 / IL-10 / Interleukin-10; Catalog#10947-HNAE; NP_000563.1; Ser 19-Asn 178) and then biotinylated.
Catalog No. 50-241-0570