Learn More
Sino Biological Anti-IL-10 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10947MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL10/IL-10/Interleukin-10 (rh IL10/IL-10/Interleukin-10; Catalog#10947-HNAE; NP_000563.1; Ser 19-Asn 178). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7929