Learn More
Sino Biological Anti-IL-10 Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 10947T16B20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-10 (rh IL-10; Catalog#10947-HNAE; NP_000563.1; Ser 19-Asn 178). IL-10 specific IgG was purified by Human IL-10 affinity chromatography and then biotinylated.
Catalog No. 50-255-5027