Learn More
Sino Biological Anti-IL-10RA/IL-10R alpha Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10419MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL10RA (rh IL10RA; Catalog#10419-H08H; NP_001549.2; Met1-Asn235). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8345