Learn More
Sino Biological Human IL10RB ELISA Pair Set 5PP (Sino Biological)
Catalog No. 502371728
Encompass_Preferred
Each vial contains 110 ng of recombinant IL10RB. Reconstitute standard powder with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
Catalog No. 50-237-1728 Supplier Sino Biological Supplier No. SEK109455
Each vial contains 110 ng of recombinant IL10RB. Reconstitute standard powder with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.