Learn More
Sino Biological Anti-IL-10RB/IL-10R beta Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10945MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL10RB extracellular domain (rh IL10RB; Catalog#10945-H08H; NP_000619.3; Met 1-Ser 220). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5030