Learn More
Sino Biological Anti-IL-10RB/IL-10R beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10945T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL10RB / IL-10RB (rh IL10RB; Catalog#10945-H08H; NP_000619.3; Met 1-Ser 220). IL10RB specific IgG was purified by human IL10RB / IL-10RB affinity chromatography.
Catalog No. 50-255-5028