Learn More
Sino Biological Human IL10RB PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10945T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL10RB / IL-10RB (rh IL10RB / IL-10RB; Catalog#10945-H08H; NP_000619.3; Met1-Ser220). IL10RB / IL-10RB specific IgG was purified by Human IL10RB / IL-10RB affinity chromatography.
Catalog No. 50-237-1217