Learn More
Sino Biological Recombinant Human IL-10RB/IL-10R beta Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10945H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domainof human IL10Rβ (NP_000619.3) (Met 1-Ser 220) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7034