Learn More
BIOHIPPO Human IL11Ra Microsample
SDP
Catalog No. 5000314929
Encompass
Human IL11Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL11Ra in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL11Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL11Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL11Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14929 Supplier BIOHIPPO Supplier No. BHE15211516
Human IL11Ra Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL11Ra in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL11Ra Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL11Ra Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL11Ra biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color