Learn More
Sino Biological Recombinant Human IL-12 Protein (His Tag)
Supplier:  Sino Biological CT050H08H50
Encompass_Preferred
A DNA sequence encoding the human IL12 complex composed of IL12B (NP_002178.2) ( Met1-Ser328) and the mature form of human IL12A (P29459-1)( Arg23-Ser219) linked by a polypeptide linker was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-228-2069