Learn More
Sino Biological Recombinant Human Neurotrophin 3/NT-3 Protein
Supplier:  Sino Biological CT050H08H500
Encompass_Preferred
A DNA sequence encoding the human NT3 (P20783-1) (Tyr139-Thr257) was expressed and purified.
Catalog No. 50-224-6659