Learn More
Proteintech Group Inc Human IL12 p70 ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE0001996T
Encompass_Preferred
Interleukin-12 (IL-12), is a cytokine that is secreted by activated phagocytes and dendritic cells and that induces interferon-γproduction by natural-killer and T lymphocytes. IL-12 (referred to as 'p70') is a 70 kDa heterodimer composed of a 35 kDa subunit (IL-12A p35) and a 40 kDa subunit (IL-12B p40). IL-12 is involved in the differentiation of native T cells into Th1 cells. It is known as a T cell-stimulating factor, which can stimulate the growth and function of T cells. IL-12 plays an important role in the activities of natural killer cells and T lymphocytes. IL-12 also has anti-angiogenic activity, which means it can block the formation of new blood vessels. IL-12 has been shown to play a critical role in the pathogenesis of a variety of immune-related diseases.
Catalog No. 50-197-4000