Learn More
Sino Biological Recombinant Human IL-12 p35/IL12A Protein (hFc Tag) 1 mg
Supplier:  Sino Biological 10021H02H1
Encompass_Preferred
A DNA sequence encoding the p35 subunit of human IL12, termed as IL12A (P29459) (Met 1-Ser 219) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6946