Learn More
Sino Biological Human IL12A PAb 20ul (Sino Biological)
Supplier:  Sino Biological 10587MM05P100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL12A (rh IL12A; Catalog#10021-H08H; P29459; Met 1-Ser 219). IL12A IgG was purified by human IL12A affinity chromatography.
Catalog No. 50-237-4399