Learn More
Sino Biological Anti-IL-12 p40/IL12B Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10052MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL12B (rhIL12B; Catalog#10052-H08H; Met 1-Ser 328; NP_002178.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2225