Learn More
Sino Biological Recombinant Anti-IL-12 p40/IL12B Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10052R05420
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL12B / p40 (rh IL12B; Catalog#10052-H08H; Met 1-Ser 328; NP_002178.2).
Catalog No. 50-241-0284