Learn More
Sino Biological Anti-IL-12 p40/IL12B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10052RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL12B (rhIL12B; Catalog#10052-H08H; Met 1-Ser 328; NP_002178.2). IL12B specific IgG was purified by human IL12B affinity chromatography.
Catalog No. 50-237-3364