Learn More
Sino Biological Recombinant Human IL-23 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological CT048H27HB100
Encompass_Preferred
A DNA sequence encoding the human IL23 complex composed of human IL12B (NP_002178.2) (Met1-Ser328) and human IL23A (NP_057668.1) (Arg20-Pro189) linked by a polypeptide linker was fused with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-2609