Learn More
Sino Biological Recombinant Human IL-23 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological CT048H08HB20
Encompass_Preferred
A DNA sequence encoding the human IL23 complex composed of IL12B (NP_002178.2) (Met1-Ser328) and the mature form of human IL23A (NP_057668.1) (Arg20-Pro189) linked by a polypeptide linker was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-2610