Learn More
Sino Biological Recombinant Human IL-12 R beta 1/IL12RB1 Protein (ECD, hFc Tag)
Catalog No. 502993377
Encompass_Preferred
A DNA sequence encoding the Human IL12RB1 (NP_005526.1) (Met1-Glu540) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-3377 Supplier Sino Biological Supplier No. 11674H02H100
A DNA sequence encoding the Human IL12RB1 (NP_005526.1) (Met1-Glu540) was expressed with the Fc region of human IgG1 at the C-terminus.