Learn More
Sino Biological Anti-IL-12 R beta 1/IL12RB1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11674MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL12RB1 (rh IL12RB1; Catalog#11674-H08H; NP_005526.1; Met1-Glu540). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4639