Learn More
Sino Biological Recombinant Human IL-13 Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Catalog No. 502992886
Encompass_Preferred
A DNA sequence encoding the Human IL13 (AAK53823.1) (Met1-Asn132) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2886 Supplier Sino Biological Supplier No. 10369H49HB100
A DNA sequence encoding the Human IL13 (AAK53823.1) (Met1-Asn132) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.