Learn More
Sino Biological Recombinant Human IL-13 Protein (His Tag), HPLC-verified
Catalog No. 502993656
Encompass_Preferred
A DNA sequence encoding the Human IL13 (AAK53823.1) (Met1-Asn132) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3656 Supplier Sino Biological Supplier No. 10369H08H50
A DNA sequence encoding the Human IL13 (AAK53823.1) (Met1-Asn132) was expressed with a polyhistidine tag at the C-terminus.