Learn More
Sino Biological Human IL13 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40592V08H132B20
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL13/IL-13/ALRH (rh IL13/IL-13/ALRH; Catalog#10369-H01H; AAK53823.1; Gly21-Asn132). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4477